Gene Transcripts
- Gene Symbol
- BTBD3
- Entrez Gene ID
- 22903
- Gene Name
- BTB domain containing 3
- Gene Aliases
- dJ742J24.1
- Chromosome Location
- Chr. 20: 11890723 - 11926595 on Build GRCh38
- UniGene ID
- Hs.709366
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282550.1 |
NP_001269479.1
|
3, 4 |
410 |
|
NM_001282551.1 |
NP_001269480.1
|
3, 4 |
506 |
|
NM_014962.3 |
NP_055777.1
|
2, 3 |
828 |
|
NM_181443.3 |
NP_852108.2
|
3, 4 |
591 |
|
XM_017027728.1 |
XP_016883217.1
|
4, 5 |
739 |
|
XM_017027729.1 |
XP_016883218.1
|
4, 5 |
671 |
|
GenBank mRNA |
AB023169.1 |
- |
2, 3 |
776 |
|
AK299281.1 |
- |
3, 4 |
591 |
|
AK307472.1 |
- |
2, 3 |
158 |
|
BC093677.1 |
- |
2, 3 |
299 |
|
BC101527.1 |
- |
2, 3 |
298 |
8069033 |
BC109315.1 |
- |
3, 4 |
439 |
40036410 |
BC109316.1 |
- |
3, 4 |
439 |
40036411 |
CB961606.1 |
- |
3, 4 |
512 |
|
DC314595.1 |
- |
3, 4 |
506 |
|
DC418438.1 |
- |
3, 4 |
410 |
|
View Full Product Details