Gene Transcripts

Gene Symbol
RRP12
Entrez Gene ID
23223
Gene Name
ribosomal RNA processing 12 homolog
Gene Aliases
KIAA0690
Chromosome Location
Chr. 10: 97356701 - 97401370 on Build GRCh38
UniGene ID
Hs.434251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145114.1 NP_001138586.1 5 759
NM_001284337.1 NP_001271266.1 4 642
NM_015179.3 NP_055994.2 7 942
XM_011539555.1 XP_011537857.1 6 811
XM_011539556.2 XP_011537858.1 7 912
GenBank mRNA AB014590.1 - 4 555
AK000238.1 - 4 352
AK022496.1 - 7 913
AK296315.1 - 5 623
BC012745.2 - 7 880 4025242

View Full Product Details