Gene Transcripts
- Gene Symbol
- NCSTN
- Entrez Gene ID
- 23385
- Gene Name
- nicastrin
- Gene Aliases
- ATAG1874
- Chromosome Location
- Chr. 1: 160343056 - 160358952 on Build GRCh38
- UniGene ID
- Hs.517249
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290184.1 |
NP_001277113.1
|
16, 17 |
2062 |
|
NM_001290186.1 |
NP_001277115.1
|
12, 13 |
1509 |
|
NM_015331.2 |
NP_056146.1
|
15, 16 |
1923 |
|
GenBank mRNA |
AF240468.1 |
- |
15, 16 |
1941 |
|
AK296153.1 |
- |
10, 11 |
1208 |
|
AK299142.1 |
- |
12, 13 |
1396 |
|
AK314764.1 |
- |
15, 16 |
1830 |
|
AY359120.1 |
- |
15, 16 |
1800 |
|
BC047621.1 |
- |
16, 17 |
2019 |
4827966 |
D87442.1 |
- |
15, 16 |
1797 |
|
View Full Product Details