Gene Transcripts

Gene Symbol
SPEF1
Entrez Gene ID
25876
Gene Name
sperm flagellar 1
Gene Aliases
C20orf28, CLAMP, SPEF1A
Chromosome Location
Chr. 20: 3777504 - 3781466 on Build GRCh38
UniGene ID
Hs.72620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015417.4 NP_056232.2 2 299
XM_005260683.4 XP_005260740.1 2 310
GenBank mRNA AK313766.1 - 2 312
AL080154.1 - 2 281
BC022476.1 - 2 258 4816764
BI520334.1 - 2 298
DB030684.1 - 2 312
DB033051.1 - 2 292
EF560745.1 - 2 297

View Full Product Details