Gene Transcripts

Gene Symbol
SAMHD1
Entrez Gene ID
25939
Gene Name
SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1
Gene Aliases
CHBL2, DCIP, HDDC1, MOP-5, SBBI88
Chromosome Location
Chr. 20: 36890882 - 36951843 on Build GRCh38
UniGene ID
Hs.580681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015474.3 NP_056289.2 4 662
XM_005260384.3 XP_005260441.1 4 635
XM_011528761.2 XP_011527063.1 4 635
GenBank mRNA AB013847.1 - 4 462
AB208944.1 - 4 492
AF228421.1 - 4 615
AK027811.1 - 4 635
AK304187.1 - 4 419
AK304795.1 - 5 590
AK315169.1 - 4 595
AL050267.1 - 4 537
BC036450.2 - 4 490 4816438

View Full Product Details