Gene Transcripts

Gene Symbol
C1QA
Entrez Gene ID
712
Gene Name
complement C1q A chain
Gene Aliases
Chromosome Location
Chr. 1: 22636625 - 22639682 on Build GRCh38
UniGene ID
Hs.632379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015991.2 NP_057075.1 3 277
GenBank mRNA AF135157.1 - 2 204
AK311980.1 - 3 220
BC030153.2 - 3 247 4850418
BC071986.1 - 2 213 6300836
BG548439.1 - 3 278

View Full Product Details