Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300779.1 | NP_001287708.1 | 3 | 429 | |
NM_015954.3 | NP_057038.2 | 3 | 429 | ||
GenBank mRNA | AF132960.1 | - | 3 | 290 | |
AK024614.1 | - | 3 | 245 | ||
AK222541.1 | - | 3 | 245 | ||
AK296697.1 | - | 3 | 186 | ||
AK309182.1 | - | 3 | 345 | ||
BC041328.1 | - | 3 | 429 | ||
BC056234.1 | - | 3 | 271 | ||
BU928832.1 | - | 3 | 235 |