Gene Transcripts

Gene Symbol
MEMO1
Entrez Gene ID
51072
Gene Name
mediator of cell motility 1
Gene Aliases
C2orf4, CGI-27, MEMO, NS5ATP7
Chromosome Location
Chr. 2: 31867188 - 32011052 on Build GRCh38
UniGene ID
Hs.444969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301833.1 NP_001288762.1 4 287
NM_001301852.1 NP_001288781.1 3 209
NM_015955.3 NP_057039.1 3 576
NR_126032.1 3 576
NR_126034.1 3 209
XM_011532892.2 XP_011531194.1 4 224
XM_011532896.2 XP_011531198.1 2 102
XM_017004255.1 XP_016859744.1 4 197
GenBank mRNA AB041018.1 - 3 576
AB041019.1 - 3 225
AF132961.1 - 3 274
AF363446.1 - 3 493
AF529368.1 - 3 159
AK057760.1 - 3 198
AK290753.1 - 4 243
BC018733.1 - 3 541 4851517
BC070046.1 - 4 316 5273070
BC094681.1 - 3 180 6205399
BG033287.1 - 3 158
BP248979.1 - 4 287
CD250945.1 - 3 273

View Full Product Details