Gene Transcripts

Gene Symbol
FCF1
Entrez Gene ID
51077
Gene Name
FCF1 rRNA-processing protein
Gene Aliases
Bka, C14orf111, CGI-35, UTP24
Chromosome Location
Chr. 14: 74713118 - 74738620 on Build GRCh38
UniGene ID
Hs.579828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318508.1 NP_001305437.1 3 209
NM_015962.4 NP_057046.1 3 173
XM_005267730.3 XP_005267787.1 3 175
GenBank mRNA AF132969.1 - 2 122
AK290666.1 - 3 154
BC080600.1 - 3 133 4889846
BX248271.1 - 3 135
BX489015.1 - 3 175
BX647810.1 - 3 199

View Full Product Details