Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
50865
Gene Name
heme binding protein 1
Gene Aliases
HBP, HEBP
Chromosome Location
Chr. 12: 12974864 - 13000309 on Build GRCh38
UniGene ID
Hs.642618
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015987.4 NP_057071.2 4 1191
GenBank mRNA BC016277.2 - 4 1026

View Full Product Details