Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017006575.1 | XP_016862064.1 | 3 | 364 | |
XM_017006578.1 | XP_016862067.1 | 3 | 344 | ||
XM_017006579.1 | XP_016862068.1 | 5 | 469 | ||
GenBank mRNA | AF114816.1 | - | 5 | 586 | |
AF114817.1 | - | 5 | 586 | ||
AK298782.1 | - | 3 | 280 |