Gene Transcripts
- Gene Symbol
- MAOB
- Entrez Gene ID
- 4129
- Gene Name
- monoamine oxidase B
- Gene Aliases
- Chromosome Location
- Chr. X: 43766610 - 43882475 on Build GRCh38
- UniGene ID
- Hs.654473
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000898.4 |
NP_000889.3
|
2, 3 |
309 |
|
XM_005272608.3 |
XP_005272665.1
|
4, 5 |
409 |
|
XM_017029523.1 |
XP_016885012.1
|
4, 5 |
459 |
|
XM_017029524.1 |
XP_016885013.1
|
3, 4 |
269 |
|
GenBank mRNA |
AK294306.1 |
- |
3, 4 |
227 |
|
AK298942.1 |
- |
2, 3 |
246 |
|
AK310458.1 |
- |
2, 3 |
281 |
|
AK312679.1 |
- |
2, 3 |
284 |
|
AU127841.1 |
- |
2, 3 |
282 |
|
BC022494.1 |
- |
2, 3 |
255 |
4800208 |
DC326844.1 |
- |
2, 3 |
310 |
|
M69177.1 |
- |
2, 3 |
210 |
|
S62734.1 |
- |
2, 3 |
133 |
|
View Full Product Details