Gene Transcripts
- Gene Symbol
- ASB3
- Entrez Gene ID
- 51130
- Gene Name
- ankyrin repeat and SOCS box containing 3
- Gene Aliases
- ASB-3
- Chromosome Location
- Chr. 2: 53669980 - 53787009 on Build GRCh38
- UniGene ID
- Hs.40763
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001201965.1 |
NP_001188894.1
|
3, 4 |
832 |
|
NM_016115.4 |
NP_057199.1
|
4, 5 |
666 |
|
NM_145863.2 |
NP_665862.1
|
3, 4 |
457 |
|
GenBank mRNA |
AB209795.1 |
- |
4, 5 |
558 |
|
AF156778.1 |
- |
3, 4 |
464 |
|
AK000985.1 |
- |
4, 5 |
592 |
|
AK001283.1 |
- |
3, 4 |
390 |
|
AK056069.1 |
- |
3, 4 |
834 |
|
AK222715.1 |
- |
3, 4 |
448 |
|
BC006488.2 |
- |
3, 4 |
478 |
3537359 |
BC009569.1 |
- |
4, 5 |
524 |
4052577 |
DA038561.1 |
- |
3, 4 |
457 |
|
- Gene Symbol
- GPR75-ASB3
- Entrez Gene ID
- 100302652
- Gene Name
- GPR75-ASB3 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 2: 53670293 - 53860033 on Build GRCh38
- UniGene ID
- Hs.40763
- Species
- Homo sapiens
View Full Product Details