Gene Transcripts

Gene Symbol
SLC45A2
Entrez Gene ID
51151
Gene Name
solute carrier family 45 member 2
Gene Aliases
1A1, AIM1, MATP, OCA4, SHEP5
Chromosome Location
Chr. 5: 33944616 - 33984675 on Build GRCh38
UniGene ID
Hs.278962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012509.3 NP_001012527.1 3 958
NM_016180.4 NP_057264.3 3 958
GenBank mRNA AF172849.1 - 3 891
BC064405.1 - 3 924 5563039

View Full Product Details