Gene Transcripts

Gene Symbol
UIMC1
Entrez Gene ID
51720
Gene Name
ubiquitin interaction motif containing 1
Gene Aliases
RAP80, X2HRIP110
Chromosome Location
Chr. 5: 176905005 - 177022640 on Build GRCh38
UniGene ID
Hs.232721
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199297.2 NP_001186226.1 3 481
NM_001199298.1 NP_001186227.1 2 256
NM_001317961.1 NP_001304890.1 2 367
NM_016290.4 NP_057374.3 2 270
XM_005265930.2 XP_005265987.1 2 244
XM_005265932.2 XP_005265989.1 2 223
XM_005265933.2 XP_005265990.1 2 335
XM_005265934.2 XP_005265991.1 2 235
XM_005265935.2 XP_005265992.1 2 244
XM_005265936.2 XP_005265993.1 2 235
XM_006714871.2 XP_006714934.1 2 219
XM_006714874.2 XP_006714937.1 2 219
XM_011534568.2 XP_011532870.1 2 235
XM_011534570.1 XP_011532872.1 2 235
XM_017009575.1 XP_016865064.1 2 244
XM_017009576.1 XP_016865065.1 2 236
XM_017009577.1 XP_016865066.1 2 288
XM_017009578.1 XP_016865067.1 2 244
XM_017009579.1 XP_016865068.1 2 144
GenBank mRNA AF349313.1 - 2 212
AK023044.1 - 3 258
AK304794.1 - 2 193
BC006078.1 - 2 177 3659756
BC032561.1 - 2 158 5578134
BP270983.1 - 2 256
BX537376.1 - 2 270
CF994701.1 - 2 380

View Full Product Details