Gene Transcripts

Gene Symbol
OR13C3
Entrez Gene ID
138803
Gene Name
olfactory receptor family 13 subfamily C member 3
Gene Aliases
OR37G, OR9-8
Chromosome Location
Chr. 9: 104535770 - 104536813 on Build GRCh38
UniGene ID
Hs.553600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001961.1 NP_001001961.1 1 880
GenBank mRNA BC136949.1 - 1 923
BC136952.1 - 1 923 9020944

View Full Product Details