Gene Transcripts
- Gene Symbol
- TRAPPC2L
- Entrez Gene ID
- 51693
- Gene Name
- trafficking protein particle complex 2-like
- Gene Aliases
- HSPC176
- Chromosome Location
- Chr. 16: 88856220 - 88862686 on Build GRCh38
- UniGene ID
- Hs.461722
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318524.1 |
NP_001305453.1
|
2, 3 |
277 |
|
NM_001318525.1 |
NP_001305454.1
|
2, 3 |
277 |
|
NM_001318528.1 |
NP_001305457.1
|
2, 3 |
254 |
|
NM_001318530.1 |
NP_001305459.1
|
2, 3 |
254 |
|
NM_016209.4 |
NP_057293.1
|
2, 3 |
277 |
|
GenBank mRNA |
AF089106.1 |
- |
2, 3 |
270 |
|
AF161524.1 |
- |
2, 3 |
232 |
|
AK126779.1 |
- |
2, 3 |
236 |
|
AK311885.1 |
- |
2, 3 |
246 |
|
AX771012.1 |
- |
2, 3 |
270 |
|
AX771013.1 |
- |
2, 3 |
232 |
|
BC011369.1 |
- |
2, 3 |
224 |
3868798 |
BC018024.1 |
- |
2, 3 |
231 |
4287920 |
BC105809.1 |
- |
2, 3 |
243 |
6745065 |
BX419849.2 |
- |
2, 3 |
222 |
|
DA209193.1 |
- |
2, 3 |
254 |
|
DA293122.1 |
- |
2, 3 |
277 |
|
DA295904.1 |
- |
2, 3 |
213 |
|
DA833474.1 |
- |
2, 3 |
216 |
|
View Full Product Details