Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256012.1 | NP_001242941.1 | 9 | 980 | |
NM_001256095.1 | NP_001243024.1 | 9 | 977 | ||
NM_005964.3 | NP_005955.3 | 8 | 950 | ||
XM_011523875.2 | XP_011522177.1 | 9 | 1269 | ||
XM_011523878.2 | XP_011522180.1 | 9 | 1321 | ||
XM_011523879.2 | XP_011522181.1 | 9 | 1266 | ||
XM_011523880.2 | XP_011522182.1 | 8 | 1239 | ||
XM_017024677.1 | XP_016880166.1 | 9 | 1035 | ||
XM_017024678.1 | XP_016880167.1 | 10 | 1017 | ||
XM_017024679.1 | XP_016880168.1 | 10 | 1058 | ||
XM_017024680.1 | XP_016880169.1 | 10 | 1366 | ||
XM_017024681.1 | XP_016880170.1 | 9 | 987 | ||
XM_017024682.1 | XP_016880171.1 | 9 | 962 | ||
GenBank mRNA | AB210026.1 | - | 9 | 932 | |
BC117690.1 | - | 7 | 852 | 40072059 | |
BC117691.1 | - | 7 | 852 | 40072061 | |
BC144668.1 | - | 9 | 934 | 9053199 | |
BC150634.1 | - | 9 | 934 | 9057004 | |
BG421687.1 | - | 4 | 221 | ||
M69181.1 | - | 8 | 890 | ||
U15618.1 | - | 5 | 226 |