Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001131028.1 | NP_001124500.1 | 3 | 507 | |
NM_031482.4 | NP_113670.1 | 2 | 372 | ||
XM_005248610.4 | XP_005248667.1 | 2 | 1642 | ||
XM_005248611.4 | XP_005248668.1 | 2 | 216 | ||
XM_005248612.3 | XP_005248669.1 | 2 | 376 | ||
XM_017009944.1 | XP_016865433.1 | 3 | 511 | ||
GenBank mRNA | AF318326.1 | - | 2 | 525 | |
AK024016.1 | - | 2 | 372 | ||
AK225969.1 | - | 2 | 372 | ||
AK315752.1 | - | 2 | 372 | ||
AL136912.1 | - | 2 | 240 | ||
BC029268.1 | - | 2 | 342 | 4521833 |