Gene Transcripts
- Gene Symbol
- TLK1
- Entrez Gene ID
- 9874
- Gene Name
- tousled like kinase 1
- Gene Aliases
- PKU-beta
- Chromosome Location
- Chr. 2: 170990823 - 171231314 on Build GRCh38
- UniGene ID
- Hs.744917
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136554.1 |
NP_001130026.1
|
11, 12 |
1192 |
|
NM_001136555.1 |
NP_001130027.1
|
9, 10 |
963 |
|
NM_012290.4 |
NP_036422.3
|
11, 12 |
1633 |
|
XM_017005421.1 |
XP_016860910.1
|
12, 13 |
1742 |
|
XM_017005422.1 |
XP_016860911.1
|
10, 11 |
1554 |
|
GenBank mRNA |
AF162666.1 |
- |
11, 12 |
1314 |
|
AF246219.1 |
- |
4, 5 |
519 |
|
AK090779.1 |
- |
11, 12 |
1192 |
|
AK091975.1 |
- |
11, 12 |
1633 |
|
AK292508.1 |
- |
11, 12 |
1274 |
|
AK301857.1 |
- |
9, 10 |
963 |
|
BC032657.1 |
- |
11, 12 |
1194 |
5532877 |
BX537631.1 |
- |
4 |
1796 |
|
D50927.2 |
- |
11, 12 |
1266 |
|
View Full Product Details