Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000779.3 | NP_000770.2 | 12 | 1780 | |
NM_001099772.1 | NP_001093242.1 | 12 | 1783 | ||
NM_001319161.1 | NP_001306090.1 | 11 | 1738 | ||
NM_001319162.1 | NP_001306091.1 | 10 | 1596 | ||
NM_001319163.1 | NP_001306092.1 | 10 | 1593 | ||
NR_135003.1 | 12 | 1710 | |||
XM_011540833.1 | XP_011539135.1 | 11 | 1416 | ||
XM_017000466.1 | XP_016855955.1 | 11 | 1687 | ||
GenBank mRNA | AK225576.1 | - | 12 | 1732 | |
BC017758.1 | - | 12 | 1735 | 4693225 | |
CD013981.1 | - | 12 | 1644 | ||
J02871.1 | - | 12 | 1708 | ||
X16699.1 | - | 12 | 1695 |