Gene Transcripts
- Gene Symbol
- SPG21
- Entrez Gene ID
- 51324
- Gene Name
- spastic paraplegia 21 (autosomal recessive, Mast syndrome)
- Gene Aliases
- ACP33, BM-019, GL010, MAST
- Chromosome Location
- Chr. 15: 64963021 - 64989946 on Build GRCh38
- UniGene ID
- Hs.242458
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127889.4 |
NP_001121361.1
|
6, 7 |
693 |
|
NM_001127890.4 |
NP_001121362.1
|
5, 6 |
793 |
|
NM_016630.6 |
NP_057714.1
|
6, 7 |
874 |
|
XM_005254437.4 |
XP_005254494.1
|
7, 8 |
907 |
|
XM_017022297.1 |
XP_016877786.1
|
6, 7 |
1242 |
|
XM_017022298.1 |
XP_016877787.1
|
7, 8 |
1332 |
|
GenBank mRNA |
AF208861.1 |
- |
5, 6 |
589 |
|
AF212231.1 |
- |
6, 7 |
661 |
|
AK123725.1 |
- |
4, 5 |
1106 |
|
AK172849.1 |
- |
6, 7 |
706 |
|
AK225527.1 |
- |
6, 7 |
706 |
|
AK301362.1 |
- |
5, 6 |
758 |
|
AK312943.1 |
- |
6, 7 |
709 |
|
BC000244.1 |
- |
6, 7 |
744 |
3357388 |
BI771887.1 |
- |
5, 6 |
627 |
|
BX446863.2 |
- |
6, 7 |
824 |
|
View Full Product Details