Gene Transcripts

Gene Symbol
IL1RAPL2
Entrez Gene ID
26280
Gene Name
interleukin 1 receptor accessory protein like 2
Gene Aliases
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
Chromosome Location
Chr. X: 104566315 - 105769729 on Build GRCh38
UniGene ID
Hs.675519
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017416.1 NP_059112.1 3 1005
XM_017029403.1 XP_016884892.1 3 423
GenBank mRNA AF181285.1 - 1 167
AF212016.1 - 2 249
AF284436.1 - 3 400
AJ272208.1 - 3 1005
AJ290436.1 - 2 268
AK290196.1 - 3 1121
BC104784.1 - 3 297 8143787
BC104786.1 - 3 297 8143789

View Full Product Details