Gene Transcripts

Gene Symbol
FAM20A
Entrez Gene ID
54757
Gene Name
family with sequence similarity 20 member A
Gene Aliases
AI1G, AIGFS, FP2747
Chromosome Location
Chr. 17: 68534561 - 68600954 on Build GRCh38
UniGene ID
Hs.268874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243746.1 NP_001230675.1 5 1202
NM_017565.3 NP_060035.2 4 976
NR_027751.1 4 382
XM_006721959.2 XP_006722022.1 4 348
XM_011524918.2 XP_011523220.1 4 816
XM_017024781.1 XP_016880270.1 4 816
GenBank mRNA AB545606.1 - 4 976
AK056789.1 - 4 816
AK298071.1 - 5 844
AL133105.1 - 2 85
AY358197.1 - 4 976
BC036222.1 - 4 345
BC136686.1 - 4 865 9020676
BC136689.1 - 4 865 9020679
DB057818.1 - 4 382

View Full Product Details