Gene Transcripts

Gene Symbol
POU6F1
Entrez Gene ID
5463
Gene Name
POU class 6 homeobox 1
Gene Aliases
BRN5, MPOU, TCFB1
Chromosome Location
Chr. 12: 51186936 - 51218134 on Build GRCh38
UniGene ID
Hs.555886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002702.3 NP_002693.3 6 2348
NR_026893.1 11 2806
XM_017019507.1 XP_016874996.1 11 3993
XM_017019508.1 XP_016874997.1 12 3904
XM_017019509.1 XP_016874998.1 10 2913
XM_017019510.1 XP_016874999.1 11 3342
XM_017019511.1 XP_016875000.1 11 3511
XM_017019512.1 XP_016875001.1 11 2867
XM_017019513.1 XP_016875002.1 11 3990
XM_017019514.1 XP_016875003.1 11 3987
XM_017019515.1 XP_016875004.1 11 3333
XM_017019516.1 XP_016875005.1 11 3984
XM_017019517.1 XP_016875006.1 10 3864
XM_017019518.1 XP_016875007.1 10 3858
XM_017019519.1 XP_016875008.1 10 3818
XM_017019520.1 XP_016875009.1 9 3220
XM_017019521.1 XP_016875010.1 9 3239
XM_017019522.1 XP_016875011.1 10 2717
XM_017019523.1 XP_016875012.1 8 2424
XM_017019524.1 XP_016875013.1 10 2815
XM_017019525.1 XP_016875014.1 11 2803
XM_017019526.1 XP_016875015.1 10 2711
XM_017019527.1 XP_016875016.1 10 2730
XM_017019528.1 XP_016875017.1 11 3236
XM_017019529.1 XP_016875018.1 10 3122
XM_017019530.1 XP_016875019.1 9 3027
GenBank mRNA AK095191.1 - 11 2806
AL832881.1 - 6 2348
BC051326.1 - 6 2151 6275848
DA026332.1 - 1 152
Z21966.1 - 5 2057

View Full Product Details