Gene Transcripts

Gene Symbol
TSPYL6
Entrez Gene ID
388951
Gene Name
TSPY like 6
Gene Aliases
Chromosome Location
Chr. 2: 54253178 - 54256272 on Build GRCh38
UniGene ID
Hs.620508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003937.2 NP_001003937.2 1 2098
GenBank mRNA AK097417.1 - 1 2052
BC068576.1 - 1 2033 4374337

View Full Product Details