Gene Transcripts

Gene Symbol
LEKR1
Entrez Gene ID
389170
Gene Name
leucine, glutamate and lysine rich 1
Gene Aliases
Chromosome Location
Chr. 3: 156825228 - 157047659 on Build GRCh38
UniGene ID
Hs.478048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004316.2 NP_001004316.2 13 2385
XM_005247441.3 XP_005247498.1 13 4197
XM_006713631.3 XP_006713694.1 13 2909
XM_006713636.2 XP_006713699.1 8 1605
XM_011512803.2 XP_011511105.1 12 2609
XM_011512804.2 XP_011511106.1 13 4430
XM_011512809.1 XP_011511111.1 10 2016
XM_011512813.2 XP_011511115.1 8 1689
XM_011512814.2 XP_011511116.1 8 1597
XM_017006374.1 XP_016861863.1 9 1793
XM_017006375.1 XP_016861864.1 9 1784
GenBank mRNA AK131473.1 - 14 2694

View Full Product Details