Gene Transcripts

Gene Symbol
Ska2
Entrez Gene ID
287598
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
Fam33a, RGD1307084
Chromosome Location
Chr. 10: 74413989 - 74431922 on Build Rnor_6.0
UniGene ID
Rn.203397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009624.1 NP_001009624.1 3 174
GenBank mRNA BC088264.1 - 3 174
FQ220730.1 - 3 173
FQ229832.1 - 3 173

View Full Product Details