Gene Transcripts

Gene Symbol
Prelid1
Entrez Gene ID
290995
Gene Name
PRELI domain containing 1
Gene Aliases
Preli, RGD1308082
Chromosome Location
Chr. 17: 9834245 - 9837285 on Build Rnor_6.0
UniGene ID
Rn.6867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009636.1 NP_001009636.1 5 974
GenBank mRNA BC088284.1 - 5 974
FQ212767.1 - 5 986
FQ226295.1 - 5 988

View Full Product Details