Gene Transcripts

Gene Symbol
Strap
Entrez Gene ID
297699
Gene Name
serine/threonine kinase receptor associated protein
Gene Aliases
Chromosome Location
Chr. 4: 171730629 - 171744877 on Build Rnor_6.0
UniGene ID
Rn.228807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011969.1 NP_001011969.1 10 1397
GenBank mRNA BC083714.1 - 10 1397

View Full Product Details