Gene Transcripts

Gene Symbol
Lipg
Entrez Gene ID
291437
Gene Name
lipase, endothelial
Gene Aliases
lipase
Chromosome Location
Chr. 18: 70903528 - 70924434 on Build Rnor_6.0
UniGene ID
Rn.199051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012741.1 NP_001012759.1 9 1442
GenBank mRNA AY916123.1 - 9 1442

View Full Product Details