Gene Transcripts

Gene Symbol
Nat3
Entrez Gene ID
290681
Gene Name
N-acetyltransferase 3
Gene Aliases
Chromosome Location
Chr. 16: 23901894 - 23908738 on Build Rnor_6.0
UniGene ID
Rn.142478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013052.1 NP_001013070.1 1 147
XM_006253021.2 XP_006253083.1 2 2344

View Full Product Details