Gene Transcripts

Gene Symbol
Ggn
Entrez Gene ID
292765
Gene Name
gametogenetin
Gene Aliases
Chromosome Location
Chr. 1: 88112272 - 88116990 on Build Rnor_6.0
UniGene ID
Rn.52869
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013065.1 NP_001013083.1 1 2050
XM_006228718.2 XP_006228780.1 2 2645
XM_006228719.2 XP_006228781.1 2 2479
XM_008759123.1 XP_008757345.1 2 2805
GenBank mRNA BC081922.1 - 1 2050
BC100147.1 - 2 1794 7111711

View Full Product Details