Gene Transcripts

Gene Symbol
Ubqlnl
Entrez Gene ID
293287
Gene Name
ubiquilin-like
Gene Aliases
RGD1307202
Chromosome Location
Chr. 1: 169342005 - 169344254 on Build Rnor_6.0
UniGene ID
Rn.122594
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013899.1 NP_001013921.1 1 2040
GenBank mRNA BC083634.1 - 1 2040

View Full Product Details