Gene Transcripts

Gene Symbol
Thg1l
Entrez Gene ID
303067
Gene Name
tRNA-histidine guanylyltransferase 1-like (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 31044885 - 31052092 on Build Rnor_6.0
UniGene ID
Rn.160832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013966.1 NP_001013988.1 2, 3 385
GenBank mRNA BC087596.1 - 2, 3 385

View Full Product Details