Gene Transcripts

Gene Symbol
Rsph6a
Entrez Gene ID
292684
Gene Name
radial spoke head 6 homolog A (Chlamydomonas)
Gene Aliases
Dmwd, Hn1-ps1, Rshl1
Chromosome Location
Chr. 1: 79959536 - 79979217 on Build Rnor_6.0
UniGene ID
Rn.55304
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024748.1 NP_001019919.1 2 861
XM_008758909.1 XP_008757131.1 2 916
GenBank mRNA BC097421.1 - 2 861

View Full Product Details