Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024877.1 | NP_001020048.1 | 15 | 1301 | |
XM_006242186.2 | XP_006242248.1 | 14 | 1263 | ||
XM_006242187.2 | XP_006242249.1 | 14 | 1263 | ||
XM_006242188.2 | XP_006242250.1 | 14 | 1224 | ||
GenBank mRNA | BC097379.1 | - | 15 | 1301 |