Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr. 17: 51177419 - 51260511 on Build GRCh38
UniGene ID
Hs.656803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017643.2 NP_060113.2 12 1511
XM_005257467.4 XP_005257524.1 11 1456
XM_005257468.4 XP_005257525.1 11 1449
XM_011524922.2 XP_011523224.1 11 1458
XM_011524923.2 XP_011523225.1 11 1455
XM_011524924.2 XP_011523226.1 11 1451
XM_011524925.1 XP_011523227.1 12 1572
XM_011524926.2 XP_011523228.1 12 1676
XM_011524928.2 XP_011523230.1 11 1613
XM_011524929.2 XP_011523231.1 12 1527
XM_011524930.1 XP_011523232.1 11 1249
XM_011524931.2 XP_011523233.1 10 1083
XM_011524933.2 XP_011523235.1 13 1633
XM_011524934.2 XP_011523236.1 11 1457
XM_011524935.2 XP_011523237.1 11 1450
XM_011524936.1 XP_011523238.1 11 1249
XM_017024782.1 XP_016880271.1 12 1823
GenBank mRNA AK000062.1 - 8 843
AK124061.1 - 11 1249
BC034364.1 - 12 1482
BC101736.1 - 7 772 8069242

View Full Product Details