Gene Transcripts

Gene Symbol
Olr77
Entrez Gene ID
405907
Gene Name
olfactory receptor 77
Gene Aliases
Chromosome Location
Chr. 1: 168224284 - 168225234 on Build Rnor_6.0
UniGene ID
Rn.141909
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001287.1 NP_001001287.1 1 663

View Full Product Details