Gene Transcripts

Gene Symbol
OR4F15
Entrez Gene ID
390649
Gene Name
olfactory receptor family 4 subfamily F member 15
Gene Aliases
Chromosome Location
Chr. 15: 101818187 - 101819125 on Build GRCh38
UniGene ID
Hs.554589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001674.1 NP_001001674.1 1 61
GenBank mRNA BC137064.1 - 1 85
BC137065.1 - 1 85 9021062

View Full Product Details