Gene Transcripts

Gene Symbol
ZNF787
Entrez Gene ID
126208
Gene Name
zinc finger protein 787
Gene Aliases
TIP20
Chromosome Location
Chr. 19: 56087363 - 56121373 on Build GRCh38
UniGene ID
Hs.397153
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002836.3 NP_001002836.2 3 1557
XM_011526445.2 XP_011524747.1 3 3241
GenBank mRNA AF000560.1 - 2 1352
BC012495.1 - 4 862
BC077728.1 - 3 1459 6210543

View Full Product Details