Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr. 9: 100302075 - 100352977 on Build GRCh38
UniGene ID
Hs.494648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161584.1 NP_001155056.1 6 1549
NM_017746.3 NP_060216.2 6 1605
XM_017014848.1 XP_016870337.1 6 1094
GenBank mRNA AB060968.1 - 6 1521
AJ420436.1 - 1 30
AK000294.1 - 6 1521
AK298916.1 - 8 1128
AK302619.1 - 6 1552
AK303662.1 - 6 1045
AK304869.1 - 5 1322
AY598337.1 - 5 1390
BC030652.2 - 6 1441 5268105

View Full Product Details