Gene Transcripts

Gene Symbol
OR2M7
Entrez Gene ID
391196
Gene Name
olfactory receptor family 2 subfamily M member 7
Gene Aliases
OR1-58
Chromosome Location
Chr. 1: 248323630 - 248324568 on Build GRCh38
UniGene ID
Hs.530570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004691.1 NP_001004691.1 1 781
GenBank mRNA BC136944.1 - 1 810
BC136946.1 - 1 810 9020938

View Full Product Details