Gene Transcripts

Gene Symbol
KRTAP5-11
Entrez Gene ID
440051
Gene Name
keratin associated protein 5-11
Gene Aliases
KRTAP5-5, KRTAP5-6, KRTAP5.11
Chromosome Location
Chr. 11: 71581855 - 71582875 on Build GRCh38
UniGene ID
Hs.679617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005405.2 NP_001005405.1 1 658
GenBank mRNA AB126080.1 - 1 658
AJ628244.1 - 1 80
BC130498.1 - 1 646
BC130502.1 - 1 646

View Full Product Details