Gene Transcripts

Gene Symbol
OR6C74
Entrez Gene ID
254783
Gene Name
olfactory receptor family 6 subfamily C member 74
Gene Aliases
Chromosome Location
Chr. 12: 55244114 - 55248226 on Build GRCh38
UniGene ID
Hs.554556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005490.1 NP_001005490.1 1 284
XM_017019126.1 XP_016874615.1 2 365
GenBank mRNA BC140754.1 - 1 374

View Full Product Details