Gene Transcripts

Gene Symbol
Tdpoz2
Entrez Gene ID
399673
Gene Name
TD and POZ domain containing 2
Gene Aliases
Chromosome Location
Chr. 3: 93651542 - 93652686 on Build GRCm38
UniGene ID
Mm.377260
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007222.3 NP_001007223.2 1 1042
GenBank mRNA BC141104.1 - 1 1042

View Full Product Details