Gene Transcripts

Gene Symbol
Clec3a
Entrez Gene ID
403395
Gene Name
C-type lectin domain family 3, member a
Gene Aliases
1110019O10Rik, Clecsf1, Gm796
Chromosome Location
Chr. 8: 114418034 - 114428133 on Build GRCm38
UniGene ID
Mm.491796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007223.3 NP_001007224.1 1 129
GenBank mRNA BC147139.1 - 1 129
BC147140.1 - 1 129 9056166

View Full Product Details