Gene Transcripts

Gene Symbol
Fbxw20
Entrez Gene ID
434440
Gene Name
F-box and WD-40 domain protein 20
Gene Aliases
Fbxw12
Chromosome Location
Chr. 9: 109217432 - 109234754 on Build GRCm38
UniGene ID
Mm.450846
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008428.3 NP_001008428.1 5 494
GenBank mRNA AK135785.1 - 5 475
AK136200.1 - 5 472
BC085247.1 - 5 475 30477089

View Full Product Details