Gene Transcripts

Gene Symbol
Creb3l2
Entrez Gene ID
362339
Gene Name
cAMP responsive element binding protein 3-like 2
Gene Aliases
Ra69, SCIRR69
Chromosome Location
Chr. 4: 64869285 - 64981519 on Build Rnor_6.0
UniGene ID
Rn.23031
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012188.1 NP_001012188.1 8, 9 1378
GenBank mRNA AY546001.1 - 8, 9 1378

View Full Product Details