Gene Transcripts

Gene Symbol
Stat4
Entrez Gene ID
367264
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
Chromosome Location
Chr. 9: 54340649 - 54457753 on Build Rnor_6.0
UniGene ID
Rn.137580
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012226.1 NP_001012226.1 11, 12 1565
GenBank mRNA BC081938.1 - 11, 12 1565

View Full Product Details